siRNA Details
| Gene Symbol: Slc5a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053521.1 | NP_445973.1 | 9 | 2029 | ||
| XM_006244251.2 | XP_006244313.1 | 9 | 2132 | |||
| XM_006244253.2 | XP_006244315.1 | 7 | 1676 | |||
| XM_008766774.1 | XP_008764996.1 | 9 | 2180 | |||
| XM_008766775.1 | XP_008764997.1 | 9 | 2129 | |||
| GenBank mRNA | AB030947.1 | - | 9 | 2029 | ||

Slc5a7
Genomic Map