siRNA Details
| Gene Symbol: Cpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053526.2 | NP_445978.1 | 2 | 433 | ||
| GenBank mRNA | AB042599.1 | - | 2 | 442 | ||
| BC088124.1 | - | 2 | 433 | 7326031 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpn1| Gene Symbol: Cpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053526.2 | NP_445978.1 | 2 | 433 | ||
| GenBank mRNA | AB042599.1 | - | 2 | 442 | ||
| BC088124.1 | - | 2 | 433 | 7326031 | ||