siRNA Details
| Gene Symbol: Slc22a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053537.2 | NP_445989.2 | 1 | 192 | ||
| GenBank mRNA | AY816233.1 | - | 1 | 192 | ||
| L27651.1 | - | 1 | 252 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a7| Gene Symbol: Slc22a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053537.2 | NP_445989.2 | 1 | 192 | ||
| GenBank mRNA | AY816233.1 | - | 1 | 192 | ||
| L27651.1 | - | 1 | 252 | |||