siRNA Details
Gene Symbol: Gna15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053542.1 | NP_445994.1 | 7 | 1360 | ||
GenBank mRNA | AB015308.1 | - | 7 | 1435 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gna15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053542.1 | NP_445994.1 | 7 | 1360 | ||
GenBank mRNA | AB015308.1 | - | 7 | 1435 |