siRNA Details
Gene Symbol: Picalm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053554.2 | NP_446006.1 | 17 | 1695 | ||
XM_006229669.2 | XP_006229731.1 | 17 | 2001 | |||
XM_006229670.2 | XP_006229732.1 | 17 | 1986 | |||
XM_006229671.2 | XP_006229733.1 | 17 | 1981 | |||
XM_006229672.2 | XP_006229734.1 | 17 | 2001 | |||
XM_006229673.2 | XP_006229735.1 | 17 | 1986 | |||
XM_006229674.2 | XP_006229736.1 | 17 | 1980 | |||
XM_006229675.2 | XP_006229737.1 | 17 | 1965 | |||
XM_006229676.2 | XP_006229738.1 | 17 | 2000 | |||
XM_006229677.2 | XP_006229739.1 | 17 | 2000 | |||
XM_006229678.2 | XP_006229740.1 | 17 | 1985 | |||
XM_006229679.2 | XP_006229741.1 | 16 | 1850 | |||
XM_006229680.2 | XP_006229742.1 | 16 | 1835 | |||
XM_006229681.2 | XP_006229743.1 | 16 | 1850 | |||
XM_006229682.2 | XP_006229744.1 | 16 | 1835 | |||
XM_006229683.2 | XP_006229745.1 | 16 | 1850 | |||
XM_006229684.2 | XP_006229746.1 | 16 | 1835 | |||
XM_006229685.2 | XP_006229747.1 | 16 | 1835 | |||
GenBank mRNA | AF041373.1 | - | 16 | 1567 | ||
AF041374.1 | - | 17 | 1696 | |||
AY699717.1 | - | 16 | 1542 | |||
AY706215.1 | - | 16 | 1557 | |||
AY724477.1 | - | 16 | 1560 | |||
BC100142.1 | - | 17 | 1756 | 7456159 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001252521, NM_001252524, NM_001252523, NM_146194, NM_001252520, and others.
Panther Classification:
Gene Ontology Categories:
