siRNA Details
| Gene Symbol: Nap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053561.2 | NP_446013.2 | 9 | 831 | ||
| XM_006241342.1 | XP_006241404.1 | 9 | 845 | |||
| XM_006241343.2 | XP_006241405.1 | 9 | 845 | |||
| XM_006241344.2 | XP_006241406.1 | 7 | 582 | |||
| GenBank mRNA | AF062594.1 | - | 9 | 813 | ||
| CF110129.1 | - | 4 | 266 | |||

Nap1l1
Genomic Map