siRNA Details
Gene Symbol: Ftcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053567.1 | NP_446019.1 | 2 | 242 | ||
GenBank mRNA | AF079233.3 | - | 2 | 242 | ||
BC091134.1 | - | 2 | 254 | 7369085 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ftcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053567.1 | NP_446019.1 | 2 | 242 | ||
GenBank mRNA | AF079233.3 | - | 2 | 242 | ||
BC091134.1 | - | 2 | 254 | 7369085 |