siRNA Details
| Gene Symbol: Pcyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053568.1 | NP_446020.1 | 14 | 1199 | ||
| XM_006247935.2 | XP_006247997.1 | 13 | 1171 | |||
| XM_006247936.2 | XP_006247998.1 | 14 | 1522 | |||
| GenBank mRNA | AF080568.1 | - | 14 | 1199 | ||
| BC078772.1 | - | 13 | 1156 | 7111560 | ||
| FQ210594.1 | - | 2 | 73 | |||

Pcyt2
Genomic Map