siRNA Details
| Gene Symbol: Sec16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053571.1 | NP_446023.1 | 4 | 608 | ||
| XM_008769682.1 | XP_008767904.1 | 10 | 4140 | |||
| XM_008769683.1 | XP_008767905.1 | 8 | 3848 | |||
| XM_008769684.1 | XP_008767906.1 | 5 | 1372 | |||
| XM_008769685.1 | XP_008767907.1 | 7 | 3125 | |||
| XM_008769686.1 | XP_008767908.1 | 3 | 409 | |||
| GenBank mRNA | AB060653.1 | - | 4 | 608 | ||

Sec16b
Genomic Map