siRNA Details
| Gene Symbol: Ece1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053596.2 | NP_446048.2 | 19 | 2757 | ||
| XM_006239275.2 | XP_006239337.1 | 19 | 3345 | |||
| XM_006239276.2 | XP_006239338.1 | 18 | 3361 | |||
| GenBank mRNA | BC072504.1 | - | 19 | 2757 | 7099038 | |
| D29683.1 | - | 19 | 2792 | |||

Ece1
Genomic Map