siRNA Details
| Gene Symbol: Acsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053607.1 | NP_446059.1 | 2 | 236 | ||
| XM_008760551.1 | XP_008758773.1 | 2 | 374 | |||
| GenBank mRNA | AB012933.1 | - | 2 | 236 | ||
| BC072497.1 | - | 2 | 228 | 7096959 | ||

Acsl5
Genomic Map