siRNA Details
Gene Symbol: Hspb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053612.2 | NP_446064.1 | 1 | 686 | ||
GenBank mRNA | AF314540.1 | - | 1 | 368 | ||
BC061748.1 | - | 1 | 686 | 5599613 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hspb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053612.2 | NP_446064.1 | 1 | 686 | ||
GenBank mRNA | AF314540.1 | - | 1 | 368 | ||
BC061748.1 | - | 1 | 686 | 5599613 |