siRNA Details
| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053615.1 | NP_446067.1 | 3, 4 | 362 | ||
| GenBank mRNA | BC127486.1 | - | 588 | 8364080 | ||
| U77582.1 | - | 3, 4 | 362 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_146087, AK166429, BC019740, BC025439, AK166318, and others.

Csnk1a1
Genomic Map