siRNA Details
| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053617.2 | NP_446069.1 | 3 | 284 | ||
| GenBank mRNA | BC091133.1 | - | 3 | 248 | ||
| BC107447.1 | - | 3 | 277 | 7323668 | ||
| DY312674.1 | - | 3 | 284 | |||
| FQ218610.1 | - | 3 | 289 | |||
| FQ218635.1 | - | 3 | 238 | |||
| FQ218700.1 | - | 3 | 289 | |||
| FQ219505.1 | - | 1 | 70 | |||
| FQ219645.1 | - | 3 | 356 | |||

Cpb2
Genomic Map