siRNA Details
| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053617.2 | NP_446069.1 | 9 | 943 | ||
| GenBank mRNA | BC091133.1 | - | 9 | 907 | ||
| BC107447.1 | - | 9 | 936 | 7323668 | ||
| FQ210930.1 | - | 3 | 255 | |||
| FQ218610.1 | - | 9 | 949 | |||
| FQ218635.1 | - | 9 | 896 | |||
| FQ218700.1 | - | 9 | 948 | |||
| FQ219505.1 | - | 7 | 729 | |||
| FQ219645.1 | - | 9 | 1015 | |||

Cpb2
Genomic Map