siRNA Details
| Gene Symbol: Il5ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053645.1 | NP_446097.1 | 3 | 279 | ||
| XM_006236977.2 | XP_006237039.1 | 5 | 670 | |||
| GenBank mRNA | AB056101.1 | - | 3 | 279 | ||
| AF324153.1 | - | 3 | 279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il5ra| Gene Symbol: Il5ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053645.1 | NP_446097.1 | 3 | 279 | ||
| XM_006236977.2 | XP_006237039.1 | 5 | 670 | |||
| GenBank mRNA | AB056101.1 | - | 3 | 279 | ||
| AF324153.1 | - | 3 | 279 | |||