siRNA Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053646.1 | NP_446098.1 | 2 | 178 | ||
| XM_006231261.2 | XP_006231323.1 | 5 | 536 | |||
| XM_006231263.1 | XP_006231325.1 | 4 | 1114 | |||
| XM_006231267.2 | XP_006231329.1 | 4 | 1120 | |||
| XM_006231268.2 | XP_006231330.1 | 4 | 1117 | |||
| XM_006231270.1 | XP_006231332.1 | 4 | 1117 | |||
| XM_008760319.1 | XP_008758541.1 | 7 | 2956 | |||
| XM_008760320.1 | XP_008758542.1 | 4 | 397 | |||
| GenBank mRNA | AB057433.1 | - | 2 | 178 | ||

Asah2
Genomic Map