siRNA Details
| Gene Symbol: Dll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053666.1 | NP_446118.1 | 9 | 1831 | ||
| XM_006228611.2 | XP_006228673.1 | 9 | 1890 | |||
| GenBank mRNA | AF084576.1 | - | 9 | 1831 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dll3| Gene Symbol: Dll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053666.1 | NP_446118.1 | 9 | 1831 | ||
| XM_006228611.2 | XP_006228673.1 | 9 | 1890 | |||
| GenBank mRNA | AF084576.1 | - | 9 | 1831 | ||