siRNA Details
| Gene Symbol: Sh2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053669.1 | NP_446121.1 | 2 | 817 | ||
| XM_006249092.2 | XP_006249154.1 | 2 | 1569 | |||
| XM_006249093.2 | XP_006249155.1 | 2 | 1570 | |||
| XM_008769112.1 | XP_008767334.1 | 3 | 1368 | |||
| XM_008769113.1 | XP_008767335.1 | 2 | 832 | |||
| GenBank mRNA | AF095576.1 | - | 2 | 817 | ||

Sh2b2
Genomic Map