siRNA Details
Gene Symbol: Rasgrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053721.1 | NP_446173.1 | 2 | 366 | ||
GenBank mRNA | AJ276774.1 | - | 2 | 366 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rasgrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053721.1 | NP_446173.1 | 2 | 366 | ||
GenBank mRNA | AJ276774.1 | - | 2 | 366 |