siRNA Details
| Gene Symbol: Pitpnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053742.1 | NP_446194.1 | 7, 8 | 476 | ||
| XM_006249522.2 | XP_006249584.1 | 7, 8 | 520 | |||
| XM_006249523.2 | XP_006249585.1 | 7, 8 | 520 | |||
| GenBank mRNA | BC061538.1 | - | 7, 8 | 508 | 6888321 | |
| D21132.1 | - | 7, 8 | 476 | |||

Pitpnb
Genomic Map