siRNA Details
| Gene Symbol: Ubqln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053747.2 | NP_446199.2 | 2 | 426 | ||
| XM_006253551.2 | XP_006253613.1 | 2 | 474 | |||
| GenBank mRNA | BC072477.1 | - | 2 | 426 | 7103662 | |
| D87950.1 | - | 2 | 286 | |||
| FQ222353.1 | - | 2 | 453 | |||

Ubqln1
Genomic Map