siRNA Details
| Gene Symbol: Suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053752.2 | NP_446204.2 | 2, 3 | 218 | ||
| GenBank mRNA | BC061537.1 | - | 2, 3 | 222 | 6920009 | |
| FQ212668.1 | - | 2, 3 | 215 | |||
| FQ214532.1 | - | 2, 3 | 177 | |||
| FQ214959.1 | - | 2, 3 | 207 | |||
| FQ215028.1 | - | 2, 3 | 176 | |||
| FQ215469.1 | - | 2, 3 | 215 | |||
| FQ216893.1 | - | 2, 3 | 207 | |||
| FQ217244.1 | - | 2, 3 | 232 | |||
| U75394.1 | - | 2, 3 | 159 | |||

Suclg1
Genomic Map