siRNA Details
| Gene Symbol: Suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053752.2 | NP_446204.2 | 8, 9 | 1021 | ||
| XM_008762973.1 | XP_008761195.1 | 7, 8 | 934 | |||
| GenBank mRNA | BC061537.1 | - | 8, 9 | 1025 | 6920009 | |
| FQ212668.1 | - | 8, 9 | 1018 | |||
| FQ214532.1 | - | 8, 9 | 980 | |||
| FQ214959.1 | - | 8, 9 | 1010 | |||
| FQ215028.1 | - | 8, 9 | 979 | |||
| FQ215469.1 | - | 8, 9 | 1018 | |||
| FQ216893.1 | - | 8, 9 | 1011 | |||
| FQ217244.1 | - | 8, 9 | 1034 | |||

Suclg1
Genomic Map