siRNA Details
Gene Symbol: Plce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053758.2 | NP_446210.1 | 2 | 860 | ||
XM_008760360.1 | XP_008758582.1 | 2 | 824 | |||
XM_008760361.1 | XP_008758583.1 | 2 | 2439 | |||
GenBank mRNA | AF323615.1 | - | 2 | 863 |
