siRNA Details
| Gene Symbol: Cyp27b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053763.1 | NP_446215.1 | 9 | 1750 | ||
| XM_008765374.1 | XP_008763596.1 | 10 | 3622 | |||
| GenBank mRNA | AB001992.1 | - | 9 | 1750 | ||
| AF000139.1 | - | 9 | 1742 | |||

Cyp27b1
Genomic Map