siRNA Details
| Gene Symbol: Tm4sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053785.1 | NP_446237.1 | 5 | 1082 | ||
| GenBank mRNA | AF205717.1 | - | 5 | 1082 | ||
| BC091131.1 | - | 5 | 897 | 7324973 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm4sf4| Gene Symbol: Tm4sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053785.1 | NP_446237.1 | 5 | 1082 | ||
| GenBank mRNA | AF205717.1 | - | 5 | 1082 | ||
| BC091131.1 | - | 5 | 897 | 7324973 | ||