siRNA Details
| Gene Symbol: Dars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053799.2 | NP_446251.1 | 2, 3 | 129 | ||
| XM_006249684.2 | XP_006249746.1 | 2, 3 | 205 | |||
| GenBank mRNA | BC072534.1 | - | 2, 3 | 254 | 7098427 | |
| J04487.1 | - | 2, 3 | 130 | |||

Dars
Genomic Map