siRNA Details
| Gene Symbol: Map3k8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053847.1 | NP_446299.1 | 2 | 592 | ||
| XM_006254048.2 | XP_006254110.1 | 2 | 612 | |||
| GenBank mRNA | FQ217924.1 | - | 2 | 625 | ||
| M94454.1 | - | 2 | 592 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k8| Gene Symbol: Map3k8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053847.1 | NP_446299.1 | 2 | 592 | ||
| XM_006254048.2 | XP_006254110.1 | 2 | 612 | |||
| GenBank mRNA | FQ217924.1 | - | 2 | 625 | ||
| M94454.1 | - | 2 | 592 | |||