siRNA Details
| Gene Symbol: Lman1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053886.2 | NP_446338.1 | 11, 12 | 1409 | ||
| GenBank mRNA | U44129.1 | - | 11, 12 | 1409 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lman1| Gene Symbol: Lman1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053886.2 | NP_446338.1 | 11, 12 | 1409 | ||
| GenBank mRNA | U44129.1 | - | 11, 12 | 1409 | ||