siRNA Details
Gene Symbol: Myt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053888.1 | NP_446340.1 | 9 | 1002 | ||
GenBank mRNA | U48809.1 | - | 9 | 1022 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
