siRNA Details
| Gene Symbol: Aldh1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053896.2 | NP_446348.2 | 2 | 209 | ||
| GenBank mRNA | BC098910.1 | - | 2 | 201 | 7455646 | |
| BE111477.1 | - | 2 | 209 | |||
| U60063.1 | - | 2 | 205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh1a2| Gene Symbol: Aldh1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053896.2 | NP_446348.2 | 2 | 209 | ||
| GenBank mRNA | BC098910.1 | - | 2 | 201 | 7455646 | |
| BE111477.1 | - | 2 | 209 | |||
| U60063.1 | - | 2 | 205 | |||