siRNA Details
| Gene Symbol: Kynu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053902.2 | NP_446354.1 | 4 | 402 | ||
| GenBank mRNA | U68168.1 | - | 4 | 403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kynu| Gene Symbol: Kynu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053902.2 | NP_446354.1 | 4 | 402 | ||
| GenBank mRNA | U68168.1 | - | 4 | 403 | ||