siRNA Details
| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053978.1 | NP_446430.1 | 2 | 152 | ||
| XM_008770195.1 | XP_008768417.1 | 2 | 521 | |||
| XM_008770196.1 | XP_008768418.1 | 2 | 521 | |||
| XM_008770197.1 | XP_008768419.1 | 2 | 730 | |||
| XM_008770198.1 | XP_008768420.1 | 2 | 521 | |||
| GenBank mRNA | BC072512.1 | - | 2 | 276 | 7104824 | |
| X78606.1 | - | 2 | 152 | |||

Rab28
Genomic Map