siRNA Details
| Gene Symbol: Bdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053995.3 | NP_446447.2 | 4 | 383 | ||
| XM_006248478.2 | XP_006248540.1 | 4 | 439 | |||
| XM_008768691.1 | XP_008766913.1 | 3 | 221 | |||
| GenBank mRNA | BC085916.1 | - | 4 | 383 | 7108483 | |

Bdh1
Genomic Map