siRNA Details
| Gene Symbol: TPX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012112.4 | NP_036244.2 | 3 | 782 | ||
| XM_011528697.2 | XP_011526999.1 | 3 | 340 | |||
| XM_011528699.2 | XP_011527001.1 | 2 | 528 | |||
| XM_011528700.2 | XP_011527002.1 | 2 | 261 | |||
| GenBank mRNA | AB024704.1 | - | 3 | 555 | ||
| AB027467.1 | - | 2 | 191 | |||
| AF098158.1 | - | 3 | 575 | |||
| AF146731.1 | - | 2 | 188 | |||
| AF287265.1 | - | 3 | 548 | |||
| AK312549.1 | - | 3 | 560 | |||
| AL117534.1 | - | 1 | 26 | |||
| AY734680.1 | - | 1 | 84 | |||
| BC004136.1 | - | 3 | 512 | 3509275 | ||
| BC020207.1 | - | 3 | 541 | 5015279 | ||
| BU181208.1 | - | 3 | 624 | |||

TPX2
Genomic Map