siRNA Details
Gene Symbol: Acsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057107.1 | NP_476448.1 | 1 | 210 | ||
GenBank mRNA | D30666.1 | - | 1 | 210 | ||
FQ221868.1 | - | 420 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Acsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057107.1 | NP_476448.1 | 1 | 210 | ||
GenBank mRNA | D30666.1 | - | 1 | 210 | ||
FQ221868.1 | - | 420 |