siRNA Details
| Gene Symbol: Pex6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057125.1 | NP_476466.1 | 17 | 2935 | ||
| GenBank mRNA | D63673.1 | - | 17 | 2935 | ||
| FQ210748.1 | - | 5 | 504 | |||
| Gene Symbol: LOC100911599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244507.1 | XP_006244569.1 | 1511 | |||

Pex6
Genomic Map