siRNA Details
| Gene Symbol: Hnrnpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057141.1 | NP_476482.1 | 7 | 443 | ||
| XM_006253552.2 | XP_006253614.1 | 9 | 850 | |||
| XM_006253553.2 | XP_006253615.1 | 8 | 546 | |||
| XM_006253554.2 | XP_006253616.1 | 9 | 850 | |||
| XM_006253555.2 | XP_006253617.1 | 8 | 778 | |||
| GenBank mRNA | BC061867.1 | - | 8 | 530 | 5598872 | |
| D17711.1 | - | 7 | 443 | |||

Hnrnpk
Genomic Map