siRNA Details
| Gene Symbol: Csrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057144.1 | NP_476485.1 | 1 | 133 | ||
| XM_006229237.2 | XP_006229299.1 | 2 | 237 | |||
| GenBank mRNA | X81193.1 | - | 1 | 133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csrp3| Gene Symbol: Csrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057144.1 | NP_476485.1 | 1 | 133 | ||
| XM_006229237.2 | XP_006229299.1 | 2 | 237 | |||
| GenBank mRNA | X81193.1 | - | 1 | 133 | ||