siRNA Details
| Gene Symbol: Folh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057185.2 | NP_476533.1 | 3 | 361 | ||
| GenBank mRNA | AF040256.1 | - | 3 | 367 | ||
| AF513486.2 | - | 3 | 361 | |||
| U75973.1 | - | 3 | 318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Folh1| Gene Symbol: Folh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057185.2 | NP_476533.1 | 3 | 361 | ||
| GenBank mRNA | AF040256.1 | - | 3 | 367 | ||
| AF513486.2 | - | 3 | 361 | |||
| U75973.1 | - | 3 | 318 | |||