siRNA Details
Gene Symbol: CBX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127321.1 | NP_001120793.1 | 2 | 224 | ||
NM_001127322.1 | NP_001120794.1 | 2 | 422 | |||
NM_012117.2 | NP_036249.1 | 2 | 270 | |||
GenBank mRNA | AK313506.1 | - | 2 | 231 | ||
BC006821.1 | - | 2 | 172 | 3448801 | ||
BP236173.1 | - | 2 | 224 | |||
BP336150.1 | - | 2 | 270 | |||
CR457418.1 | - | 1 | 104 | |||
DT217255.1 | - | 2 | 422 | |||
EU794599.1 | - | 2 | 239 | |||
L07515.1 | - | 2 | 239 | |||
S62077.1 | - | 2 | 238 | |||
U26311.1 | - | 1 | 102 |
