siRNA Details
| Gene Symbol: Hadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057186.2 | NP_476534.1 | 8 | 1211 | ||
| GenBank mRNA | AF095449.1 | - | 8 | 1211 | ||
| FQ212781.1 | - | 8 | 1192 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hadh| Gene Symbol: Hadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057186.2 | NP_476534.1 | 8 | 1211 | ||
| GenBank mRNA | AF095449.1 | - | 8 | 1211 | ||
| FQ212781.1 | - | 8 | 1192 | |||