siRNA Details
| Gene Symbol: Wipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057192.2 | NP_476540.2 | 6 | 1482 | ||
| GenBank mRNA | AJ303456.1 | - | 5 | 1315 | ||
| BC072475.1 | - | 6 | 1482 | 7100751 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wipf1| Gene Symbol: Wipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057192.2 | NP_476540.2 | 6 | 1482 | ||
| GenBank mRNA | AJ303456.1 | - | 5 | 1315 | ||
| BC072475.1 | - | 6 | 1482 | 7100751 | ||