siRNA Details
| Gene Symbol: Il10ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057193.2 | NP_476541.2 | 2 | 209 | ||
| XM_006242864.2 | XP_006242926.1 | 2 | 387 | |||
| GenBank mRNA | AJ305049.1 | - | 2 | 209 | ||
| FQ223464.1 | - | 2 | 230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il10ra| Gene Symbol: Il10ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057193.2 | NP_476541.2 | 2 | 209 | ||
| XM_006242864.2 | XP_006242926.1 | 2 | 387 | |||
| GenBank mRNA | AJ305049.1 | - | 2 | 209 | ||
| FQ223464.1 | - | 2 | 230 | |||