siRNA Details
| Gene Symbol: Uba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057205.2 | NP_476553.1 | 3, 4 | 188 | ||
| XM_006236936.2 | XP_006236998.1 | 2, 3 | 171 | |||
| GenBank mRNA | AF336829.1 | - | 3, 4 | 197 | ||
| BC081743.1 | - | 3, 4 | 188 | 7124631 | ||
| FQ234407.1 | - | 2, 3 | 147 | |||

Uba3
Genomic Map