siRNA Details
Gene Symbol: Slc8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078619.1 | NP_511174.1 | 1 | 587 | ||
GenBank mRNA | U08141.1 | - | 1 | 587 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078619.1 | NP_511174.1 | 1 | 587 | ||
GenBank mRNA | U08141.1 | - | 1 | 587 |