siRNA Details
| Gene Symbol: Pcyt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078622.2 | NP_511177.2 | 3 | 282 | ||
| XM_006248444.2 | XP_006248506.1 | 3 | 435 | |||
| XM_008768694.1 | XP_008766916.1 | 3 | 633 | |||
| GenBank mRNA | AW919077.1 | - | 3 | 283 | ||
| BC085713.1 | - | 3 | 280 | 7127059 | ||
| L13245.1 | - | 2 | 179 | |||
| M36071.1 | - | 3 | 279 | |||
| U03490.1 | - | 3 | 244 | |||

Pcyt1a
Genomic Map