siRNA Details
| Gene Symbol: Pcyt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078622.2 | NP_511177.2 | 8 | 844 | ||
| XM_006248444.2 | XP_006248506.1 | 8 | 997 | |||
| XM_008768694.1 | XP_008766916.1 | 8 | 1195 | |||
| GenBank mRNA | BC085713.1 | - | 8 | 842 | 7127059 | |
| L13245.1 | - | 7 | 741 | |||
| M36071.1 | - | 8 | 841 | |||
| U03490.1 | - | 8 | 806 | |||

Pcyt1a
Genomic Map