siRNA Details
| Gene Symbol: Apoa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277264.1 | NP_001264193.1 | 3 | 199 | ||
| NM_080576.2 | NP_542143.1 | 4 | 239 | |||
| XM_006242865.2 | XP_006242927.1 | 4 | 337 | |||
| GenBank mRNA | AF202887.1 | - | 3 | 184 | ||
| AF202888.1 | - | 3 | 184 | |||
| BC089780.1 | - | 4 | 239 | 7371817 | ||
| FQ210962.1 | - | 3 | 199 | |||
| FQ219463.1 | - | 3 | 199 | |||

Apoa5
Genomic Map